Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028639.3 | NP_082915.1 | 4 - 5 | 989 | 72 | |
GenBank mRNA | AK040578.1 | - | 4 - 5 | 992 | 72 | |
AK040610.1 | - | 4 - 5 | 960 | 72 | ||
AK050263.1 | - | 4 - 5 | 976 | 72 | ||
AK080284.1 | - | 4 - 5 | 968 | 72 | ||
AK153704.1 | - | 4 - 5 | 971 | 72 | ||
BC042512.1 | - | 4 - 5 | 666 | 4223801 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |