Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197028.1 | NP_001183957.1 | 5 - 6 | 701 | 96 | |
NM_028813.2 | NP_083089.1 | 6 - 7 | 767 | 96 | ||
GenBank mRNA | AK013193.1 | - | 6 - 7 | 678 | 96 | |
AK051606.1 | - | 5 - 6 | 592 | 96 | ||
BC034120.1 | - | 6 - 7 | 690 | 4527027 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |