Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175930.5 | NP_787126.3 | 10 - 11 | 1303 | 68 | |
GenBank mRNA | AK029995.1 | - | 10 - 11 | 1304 | 68 | |
AK030016.1 | - | 10 - 11 | 1257 | 68 | ||
AK162220.1 | - | 8 - 9 | 936 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |