Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134139.1 | NP_598900.1 | 3 - 4 | 295 | 101 | |
GenBank mRNA | AK077521.1 | - | 3 - 4 | 296 | 101 | |
AK145704.1 | - | 3 - 4 | 308 | 101 | ||
BC019968.1 | - | 3 - 4 | 295 | 4167260 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |