Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930568A12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016244.1 | - | 2 - 3 | 234 | 87 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY715855.1 | - | 232 | Chr. - - [-] on Build | 87 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
protein tyrosine phosphatase, non-receptor type 18 |
Ptpn18 |
FLP1, HSCF, PTP-HSCF, PTP-K1, Ptpk1 |
-6.8kb | Anti-sense | Chr. 1 - 34459746 - 34473784 on NCBI Build GRCm38 |
Downstream |
protease, serine 39 |
Prss39 | Tesp1 | 13.5kb | Anti-sense | Chr. 1 - 34498430 - 34503062 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |