Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054056.2 | NP_473397.1 | 3 - 4 | 816 | 64 | |
GenBank mRNA | AF377871.1 | - | 2 - 3 | 219 | 64 | |
DQ363525.1 | - | 3 - 4 | 816 | 64 | ||
DQ449074.1 | - | 2 - 3 | 627 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |