Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178711.3 | NP_848826.1 | 6 - 7 | 1184 | 73 | |
GenBank mRNA | AK035919.1 | - | 6 - 7 | 1185 | 73 | |
AK054258.1 | - | 6 - 7 | 742 | 73 | ||
AK082330.1 | - | 5 - 6 | 627 | 73 | ||
AK141580.1 | - | 710 | 73 | |||
AK165919.1 | - | 6 - 7 | 1185 | 73 | ||
AK166080.1 | - | 6 - 7 | 676 | 73 | ||
BC052067.1 | - | 6 - 7 | 738 | 5708712 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |