Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019570.3 | NP_062516.2 | 10 - 11 | 1880 | 79 | |
GenBank mRNA | AB057418.1 | - | 9 - 10 | 1669 | 79 | |
AK048838.1 | - | 10 - 11 | 1885 | 79 | ||
BC006721.1 | - | 5 - 6 | 1204 | 79 | ||
BC058093.1 | - | 10 - 11 | 1880 | 6827108 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |