Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080549.2 | NP_001074018.1 | 6 - 7 | 1775 | 107 | |
NM_001290459.1 | NP_001277388.1 | 5 - 6 | 1628 | 107 | ||
GenBank mRNA | AK083436.1 | - | 5 - 6 | 1616 | 107 | |
BC116300.1 | - | 6 - 7 | 1718 | 40095063 | 107 | |
BC116301.1 | - | 5 - 6 | 1571 | 40095064 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |