Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008922.2 | NP_032948.1 | 10 - 11 | 1193 | 81 | |
GenBank mRNA | AK076095.1 | - | 10 - 11 | 1175 | 81 | |
AK077760.1 | - | 10 - 11 | 1180 | 81 | ||
AK153970.1 | - | 10 - 11 | 1154 | 81 | ||
AK166687.1 | - | 4 - 5 | 399 | 81 | ||
AK168246.1 | - | 10 - 11 | 1186 | 81 | ||
BC019500.1 | - | 10 - 11 | 1154 | 3602929 | 81 | |
D13545.1 | - | 10 - 11 | 1157 | 81 | ||
D17385.1 | - | 10 - 11 | 1137 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |