Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178907.3 | NP_849238.1 | 5 - 6 | 916 | 78 | |
GenBank mRNA | AK051962.1 | - | 4 - 5 | 1924 | 78 | |
AK087496.1 | - | 4 - 5 | 577 | 78 | ||
AK144637.1 | - | 5 - 6 | 854 | 78 | ||
AK151881.1 | - | 5 - 6 | 839 | 78 | ||
AK170456.1 | - | 5 - 6 | 912 | 78 | ||
AK172344.1 | - | 5 - 6 | 827 | 78 | ||
AK172578.1 | - | 5 - 6 | 819 | 78 | ||
BC031467.1 | - | 5 - 6 | 916 | 4189474 | 78 | |
DQ899946.1 | - | 4 - 5 | 511 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |