Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900025537.mm.1 |
NM_011251.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025537.mm.1 |
NM_011251.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025537.mm.1 |
NM_029169.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025537.mm.1 |
NM_029169.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025537.mm.1 |
TC0900003044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025539.mm.1 |
NM_011251.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025539.mm.1 |
NM_011251.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025539.mm.1 |
NM_029169.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025539.mm.1 |
NM_029169.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025539.mm.1 |
TC0900003044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025536.mm.1 |
NM_011251.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025536.mm.1 |
NM_011251.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025536.mm.1 |
NM_029169.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025536.mm.1 |
NM_029169.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025536.mm.1 |
TC0900003044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900025538.mm.1 |
NM_011251.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025538.mm.1 |
NM_011251.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025538.mm.1 |
NM_029169.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025538.mm.1 |
NM_029169.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900025538.mm.1 |
TC0900003044.mm.1 |
MTA TC |