Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011842.3 | NP_035972.3 | 7 - 8 | 1029 | 60 | |
GenBank mRNA | AK077548.1 | - | 7 - 8 | 1011 | 60 | |
AK088158.1 | - | 7 - 8 | 942 | 60 | ||
AK147099.1 | - | 7 - 8 | 1008 | 60 | ||
AK149834.1 | - | 7 - 8 | 1075 | 60 | ||
AK157717.1 | - | 7 - 8 | 933 | 60 | ||
AK159979.1 | - | 7 - 8 | 1005 | 60 | ||
BC079847.1 | - | 7 - 8 | 1029 | 6419411 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |