Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271496.1 | NP_001258425.1 | 3 - 4 | 809 | 73 | |
NR_073190.1 | 4 - 5 | 869 | 73 | |||
GenBank mRNA | AK005057.1 | - | 4 - 5 | 870 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |