Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081278.2 | NP_001074747.2 | 15 - 16 | 3716 | 77 | |
GenBank mRNA | AK047705.1 | - | 5 - 6 | 887 | 77 | |
AK171689.1 | - | 4 - 5 | 907 | 77 | ||
AK172575.1 | - | 4 - 5 | 787 | 77 | ||
AK220347.1 | - | 15 - 16 | 2126 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |