Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_001114595.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_001114595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_001114596.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_001114596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_001114597.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_001114597.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_053252.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
NM_053252.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008829.mm.1 |
TC1900000980.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008828.mm.1 |
NM_001114596.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008828.mm.1 |
NM_001114596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008828.mm.1 |
TC1900000980.mm.1 |
MTA TC |