Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025572.1 | NP_001020743.1 | 10 - 11 | 5909 | 118 | |
GenBank mRNA | AK035216.1 | - | 2 - 3 | 291 | 118 | |
AK045925.1 | - | 2 - 3 | 944 | 118 | ||
AK122392.1 | - | 2 - 3 | 2270 | 118 | ||
BC028980.1 | - | 2 - 3 | 279 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |