Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198949.1 | NP_001185878.1 | 5 - 6 | 1517 | 68 | |
NM_009067.5 | NP_033093.2 | 5 - 6 | 1841 | 68 | ||
GenBank mRNA | AK136252.1 | - | 3 - 4 | 907 | 68 | |
AK145387.1 | - | 5 - 6 | 1381 | 68 | ||
AK159404.1 | - | 5 - 6 | 1394 | 68 | ||
BC054329.1 | - | 5 - 6 | 1432 | 68 | ||
BC067073.1 | - | 4 - 5 | 1276 | 30611880 | 68 | |
BC075732.1 | - | 5 - 6 | 1466 | 6507517 | 68 | |
BC076636.1 | - | 5 - 6 | 1433 | 30658617 | 68 | |
BC138546.1 | - | 4 - 5 | 1282 | 8861566 | 68 | |
BC145815.1 | - | 4 - 5 | 1282 | 40131118 | 68 | |
X80937.1 | - | 5 - 6 | 1418 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |