Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029896.1 | NP_084172.1 | 5 - 6 | 834 | 93 | |
GenBank mRNA | AK035109.1 | - | 5 - 6 | 836 | 93 | |
AK037620.1 | - | 5 - 6 | 841 | 93 | ||
AK149692.1 | - | 5 - 6 | 835 | 93 | ||
AK159240.1 | - | 5 - 6 | 834 | 93 | ||
BC019115.1 | - | 4 - 5 | 360 | 93 | ||
BC031502.1 | - | 5 - 6 | 524 | 93 | ||
BC094419.1 | - | 5 - 6 | 797 | 93 | ||
BC157943.1 | - | 757 | 9007278 | 93 | ||
BC157945.1 | - | 758 | 9007280 | 93 | ||
BC158054.1 | - | 758 | 9007356 | 93 | ||
EF011615.1 | - | 5 - 6 | 523 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |