Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080556.3 | NP_542123.3 | 6 - 7 | 904 | 72 | |
GenBank mRNA | AK010325.1 | - | 6 - 7 | 861 | 72 | |
AK030340.1 | - | 6 - 7 | 834 | 73 | ||
AK048012.1 | - | 6 - 7 | 823 | 72 | ||
AK050338.1 | - | 6 - 7 | 851 | 72 | ||
AK075720.1 | - | 6 - 7 | 904 | 72 | ||
AK088905.1 | - | 6 - 7 | 876 | 72 | ||
AK159757.1 | - | 6 - 7 | 849 | 72 | ||
AK212229.1 | - | 2 - 3 | 239 | 72 | ||
BC003862.1 | - | 6 - 7 | 843 | 3488326 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |