Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027864.2 | NP_082140.2 | 8 - 9 | 1219 | 72 | |
GenBank mRNA | AB175681.1 | - | 8 - 9 | 827 | 72 | |
AK002747.1 | - | 8 - 9 | 1191 | 72 | ||
AK049598.1 | - | 8 - 9 | 1165 | 72 | ||
AK078292.1 | - | 8 - 9 | 832 | 72 | ||
BC037716.1 | - | 2 - 3 | 198 | 72 | ||
BC117800.1 | - | 8 - 9 | 1003 | 40092267 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |