Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009045.4 | NP_033071.1 | 1 - 2 | 351 | 112 | |
GenBank mRNA | AK089489.1 | - | 1 - 2 | 96 | 112 | |
AK144565.1 | - | 1 - 2 | 96 | 112 | ||
AK154630.1 | - | 1 - 2 | 96 | 112 | ||
AK155321.1 | - | 1 - 2 | 362 | 112 | ||
AK155889.1 | - | 1 - 2 | 93 | 112 | ||
AK157076.1 | - | 1 - 2 | 83 | 112 | ||
AK158072.1 | - | 1 - 2 | 361 | 112 | ||
BC003818.1 | - | 1 - 2 | 110 | 3489295 | 112 | |
BC094053.1 | - | 1 - 2 | 351 | 3157927 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |