Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009045.4 | NP_033071.1 | 6 - 7 | 908 | 74 | |
GenBank mRNA | AK089489.1 | - | 6 - 7 | 653 | 74 | |
AK144565.1 | - | 6 - 7 | 653 | 74 | ||
AK154630.1 | - | 6 - 7 | 653 | 74 | ||
AK155889.1 | - | 6 - 7 | 650 | 74 | ||
AK157076.1 | - | 6 - 7 | 640 | 74 | ||
AK158072.1 | - | 6 - 7 | 918 | 74 | ||
BC003818.1 | - | 6 - 7 | 667 | 3489295 | 74 | |
BC094053.1 | - | 6 - 7 | 908 | 3157927 | 74 | |
DQ020177.1 | - | 4 - 5 | 477 | 74 | ||
M61909.1 | - | 6 - 7 | 623 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |