Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110796.2 | NP_001104266.1 | 7 - 8 | 13392 | 144 | |
NM_011995.4 | NP_036125.4 | 7 - 8 | 13392 | 144 | ||
GenBank mRNA | AB083477.1 | - | 7 - 8 | 13635 | 144 | |
AB083478.1 | - | 7 - 8 | 13635 | 144 | ||
Y19185.2 | - | 8 - 9 | 13302 | 144 | ||
Y19186.2 | - | 8 - 9 | 13302 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |