Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290654.1 | NP_001277583.1 | 1 - 2 | 268 | 60 | |
GenBank mRNA | BC038397.1 | - | 1 - 2 | 159 | 3676293 | 60 |
BY023007.1 | - | 1 - 2 | 157 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |