Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172256.1 | NP_758460.1 | 11 - 12 | 978 | 82 | |
GenBank mRNA | AK008822.1 | - | 10 - 11 | 928 | 82 | |
AK160259.1 | - | 11 - 12 | 992 | 82 | ||
BC030450.1 | - | 11 - 12 | 980 | 5400492 | 82 | |
BC039070.1 | - | 11 - 12 | 978 | 3482503 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |