Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckipsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030729.4 | NP_109654.2 | 8 - 9 | 1568 | 59 | |
GenBank mRNA | AF130313.2 | - | 8 - 9 | 1464 | 59 | |
AK134725.1 | - | 8 - 9 | 1550 | 59 | ||
BC064818.1 | - | 8 - 9 | 1568 | 30292108 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |