Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011830.3 | NP_035960.2 | 3 - 4 | 280 | 70 | |
GenBank mRNA | AK002676.1 | - | 3 - 4 | 312 | 70 | |
AK011777.1 | - | 3 - 4 | 313 | 70 | ||
AK143505.1 | - | 3 - 4 | 321 | 70 | ||
AK150713.1 | - | 3 - 4 | 316 | 70 | ||
AK151233.1 | - | 3 - 4 | 316 | 70 | ||
AK151706.1 | - | 3 - 4 | 339 | 70 | ||
AK152691.1 | - | 3 - 4 | 313 | 70 | ||
AK159822.1 | - | 3 - 4 | 313 | 70 | ||
AK160569.1 | - | 3 - 4 | 313 | 70 | ||
AK166667.1 | - | 3 - 4 | 315 | 70 | ||
AK169129.1 | - | 3 - 4 | 346 | 70 | ||
AK169788.1 | - | 3 - 4 | 346 | 70 | ||
AK215538.1 | - | 1 - 2 | 93 | 70 | ||
BC010314.1 | - | 3 - 4 | 280 | 2646679 | 70 | |
BC052671.1 | - | 3 - 4 | 301 | 30058399 | 70 | |
M33934.1 | - | 3 - 4 | 313 | 70 | ||
M98333.1 | - | 3 - 4 | 247 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |