Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028719.1 | NP_082995.1 | 12 - 13 | 1425 | 77 | |
GenBank mRNA | AK014396.1 | - | 9 - 10 | 1181 | 77 | |
AK043692.1 | - | 12 - 13 | 1438 | 77 | ||
BC043087.1 | - | 12 - 13 | 1425 | 6417611 | 77 | |
BC063081.1 | - | 12 - 13 | 1425 | 6417611 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |