Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esyt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177775.3 | NP_808443.2 | 18 - 19 | 2395 | 62 | |
GenBank mRNA | AK086367.1 | - | 18 - 19 | 2396 | 62 | |
AK220532.1 | - | 18 - 19 | 2383 | 62 | ||
BC118967.2 | - | 2343 | 40097262 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |