Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030133.3 | NP_084409.3 | 18 - 19 | 2369 | 70 | |
GenBank mRNA | AK144490.1 | - | 2 - 3 | 218 | 70 | |
BC038140.1 | - | 2 - 3 | 313 | 70 | ||
BC100359.1 | - | 11 - 12 | 1233 | 70 | ||
BC147429.1 | - | 2 - 3 | 246 | 9056482 | 70 | |
BC147430.1 | - | 2 - 3 | 246 | 9056479 | 70 | |
BC172066.1 | - | 3 - 4 | 353 | 9054352 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |