Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163505.1 | NP_001156977.1 | 8 - 9 | 1122 | 137 | |
NM_146091.4 | NP_666203.3 | 8 - 9 | 1024 | 137 | ||
GenBank mRNA | AK028842.1 | - | 8 - 9 | 885 | 137 | |
AK030660.1 | - | 8 - 9 | 884 | 137 | ||
AK077752.1 | - | 8 - 9 | 887 | 137 | ||
AK147720.1 | - | 8 - 9 | 982 | 137 | ||
AK169476.1 | - | 8 - 9 | 884 | 137 | ||
BC002149.1 | - | 5 - 6 | 414 | 137 | ||
BC017138.1 | - | 8 - 9 | 873 | 4485115 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |