Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177438.1 | NP_001170909.1 | 8 - 9 | 1397 | 60 | |
GenBank mRNA | AK048310.1 | - | 258 | 60 | ||
AK136505.1 | - | 8 - 9 | 1399 | 60 | ||
BC100370.1 | - | 255 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |