Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011224.1 | NP_035354.1 | 15 - 16 | 1876 | 88 | |
GenBank mRNA | AF124787.1 | - | 15 - 16 | 1876 | 88 | |
BC012961.1 | - | 15 - 16 | 1882 | 3989941 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |