Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027908.1 | NP_082184.1 | 4 - 5 | 135 | ||
GenBank mRNA | AK005066.1 | - | 4 - 5 | 135 | ||
AK090143.1 | - | 4 - 5 | 135 | |||
BC022626.1 | - | 4 - 5 | 533 | 135 | ||
BC043681.1 | - | 4 - 5 | 588 | 135 | ||
BC055025.1 | - | 4 - 5 | 553 | 135 | ||
BC118616.1 | - | 4 - 5 | 642 | 40107296 | 135 | |
BC121826.1 | - | 4 - 5 | 641 | 40107295 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |