Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164885.1 | NP_001158357.1 | 12 - 13 | 1943 | 81 | |
NM_022882.4 | NP_075020.2 | 12 - 13 | 1819 | 81 | ||
GenBank mRNA | AF286723.1 | - | 12 - 13 | 1819 | 81 | |
BC039698.1 | - | 12 - 13 | 1932 | 5101211 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |