Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133831.3 | NP_598592.2 | 1 | 115 | ||
GenBank mRNA | AK077341.1 | - | 1 | 115 | ||
AK088461.1 | - | 1 | 115 | |||
AK146916.1 | - | 1 | 115 | |||
AK151836.1 | - | 1 | 115 | |||
AK160070.1 | - | 1 | 115 | |||
AK169290.1 | - | 1 | 115 | |||
AY535001.1 | - | 1 | 115 | |||
BC017637.1 | - | 1 | 4912415 | 115 | ||
BC025810.1 | - | 1 | 115 | |||
BY283382.1 | - | 1 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |