Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029878.1 | NP_001025049.1 | 9 - 10 | 1211 | 69 | |
GenBank mRNA | BC139127.1 | - | 9 - 10 | 1262 | 8862149 | 69 |
BC139130.1 | - | 9 - 10 | 1262 | 8862152 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |