Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 13 - 14 | 2254 | 78 | |
GenBank mRNA | AK147671.1 | - | 13 - 14 | 2270 | 78 | |
AK172931.1 | - | 3 - 4 | 563 | 78 | ||
BC079860.1 | - | 13 - 14 | 2254 | 6834745 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |