Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001984.2 | NP_001001984.2 | 19 - 20 | 3908 | 68 | |
GenBank mRNA | AK141779.1 | - | 5 - 6 | 1149 | 68 | |
AK173084.1 | - | 8 - 9 | 1726 | 68 | ||
BC036264.1 | - | 7 - 8 | 1607 | 68 | ||
BC037117.1 | - | 12 - 13 | 2457 | 68 | ||
BC057051.1 | - | 6 - 7 | 1281 | 68 | ||
BC076576.1 | - | 19 - 20 | 3473 | 6840591 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |