Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198113.2 | NP_932781.1 | 2 - 3 | 203 | 98 | |
GenBank mRNA | AB099289.1 | - | 2 - 3 | 201 | 98 | |
AK148293.1 | - | 2 - 3 | 197 | 98 | ||
AK149880.1 | - | 2 - 3 | 212 | 98 | ||
BC028922.1 | - | 2 - 3 | 203 | 3987714 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |