Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033128.3 | NP_001028300.1 | 13 - 14 | 1367 | 54 | |
GenBank mRNA | AK037753.1 | - | 13 - 14 | 1353 | 54 | |
AK045850.1 | - | 13 - 14 | 1365 | 54 | ||
AK163815.1 | - | 4 - 5 | 535 | 54 | ||
BC076577.1 | - | 13 - 14 | 1356 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |