Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033128.3 | NP_001028300.1 | 15 - 16 | 1628 | 70 | |
GenBank mRNA | AK037753.1 | - | 15 - 16 | 1614 | 70 | |
AK045850.1 | - | 15 - 16 | 1626 | 70 | ||
AK163815.1 | - | 6 - 7 | 796 | 70 | ||
BC076577.1 | - | 15 - 16 | 1617 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |