Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133771.2 | NP_598532.1 | 2 - 3 | 257 | 64 | |
GenBank mRNA | AF363447.1 | - | 3 - 4 | 319 | 64 | |
AK053089.1 | - | 2 - 3 | 235 | 64 | ||
AK159560.1 | - | 2 - 3 | 221 | 64 | ||
BC013819.1 | - | 2 - 3 | 274 | 3489363 | 64 | |
BF466279.1 | - | 2 - 3 | 257 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |