Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133771.2 | NP_598532.1 | 4 - 5 | 439 | 112 | |
GenBank mRNA | AF363447.1 | - | 5 - 6 | 501 | 112 | |
AK159560.1 | - | 4 - 5 | 403 | 112 | ||
BC013819.1 | - | 4 - 5 | 456 | 3489363 | 112 | |
BF466279.1 | - | 4 - 5 | 439 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |