Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037757.1 | NP_001032846.1 | 4 - 5 | 496 | 63 | |
NM_026833.1 | NP_081109.1 | 4 - 5 | 513 | 63 | ||
GenBank mRNA | AJ304796.1 | - | 4 - 5 | 445 | 63 | |
AK033616.1 | - | 4 - 5 | 490 | 63 | ||
AK149967.1 | - | 4 - 5 | 429 | 63 | ||
AK154400.1 | - | 4 - 5 | 498 | 63 | ||
AK159820.1 | - | 4 - 5 | 441 | 63 | ||
AK171737.1 | - | 4 - 5 | 431 | 63 | ||
AK172673.1 | - | 4 - 5 | 431 | 63 | ||
BC025839.1 | - | 4 - 5 | 426 | 5136195 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |