Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037757.1 | NP_001032846.1 | 9 - 10 | 1318 | 75 | |
NM_026833.1 | NP_081109.1 | 9 - 10 | 1335 | 75 | ||
GenBank mRNA | AJ304796.1 | - | 9 - 10 | 1267 | 75 | |
AK004209.1 | - | 3 - 4 | 441 | 75 | ||
AK033616.1 | - | 9 - 10 | 1312 | 75 | ||
AK149967.1 | - | 9 - 10 | 1251 | 75 | ||
AK154400.1 | - | 9 - 10 | 1320 | 75 | ||
AK159820.1 | - | 9 - 10 | 1263 | 75 | ||
AK171737.1 | - | 9 - 10 | 1253 | 75 | ||
AK172673.1 | - | 9 - 10 | 1253 | 75 | ||
BC025839.1 | - | 9 - 10 | 1248 | 5136195 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |