Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007658.3 | NP_031684.3 | 8 - 9 | 1144 | 119 | |
GenBank mRNA | AK077609.1 | - | 8 - 9 | 761 | 119 | |
AK077991.1 | - | 8 - 9 | 1147 | 119 | ||
AK142334.1 | - | 7 - 8 | 1419 | 119 | ||
BC046296.1 | - | 8 - 9 | 763 | 6486343 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |