Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290181.1 | NP_001277110.1 | 2 - 3 | 2028 | 87 | |
NM_001290182.1 | NP_001277111.1 | 2 - 3 | 288 | 87 | ||
NM_024437.4 | NP_077757.2 | 2 - 3 | 288 | 87 | ||
NM_024446.5 | NP_077766.3 | 2 - 3 | 288 | 87 | ||
GenBank mRNA | AA616044.1 | - | 1 - 2 | 130 | 87 | |
AF338424.1 | - | 2 - 3 | 248 | 87 | ||
AK004924.1 | - | 2 - 3 | 1984 | 87 | ||
AK011172.1 | - | 2 - 3 | 222 | 87 | ||
BC033046.1 | - | 2 - 3 | 220 | 87 | ||
BC069843.1 | - | 2 - 3 | 244 | 5102473 | 87 | |
CA460240.1 | - | 2 - 3 | 320 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |