Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 3 - 4 | 320 | 119 | |
GenBank mRNA | AK009346.1 | - | 3 - 4 | 293 | 119 | |
AK009504.1 | - | 3 - 4 | 295 | 119 | ||
AK009619.1 | - | 3 - 4 | 293 | 119 | ||
AK009776.1 | - | 3 - 4 | 277 | 119 | ||
AK009794.1 | - | 3 - 4 | 291 | 119 | ||
AK009871.1 | - | 3 - 4 | 295 | 119 | ||
AK010130.1 | - | 3 - 4 | 277 | 119 | ||
AK010154.1 | - | 3 - 4 | 291 | 119 | ||
AK010157.1 | - | 3 - 4 | 321 | 119 | ||
AK010215.1 | - | 3 - 4 | 291 | 119 | ||
AK028158.1 | - | 3 - 4 | 294 | 119 | ||
AK075874.1 | - | 3 - 4 | 291 | 119 | ||
AK160516.1 | - | 3 - 4 | 291 | 119 | ||
BC116681.1 | - | 3 - 4 | 275 | 40106489 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |